Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BPTF SIRNA Assays ›
Gene Symbol:
BPTF
Gene Name:
bromodomain PHD finger transcription factor
Gene Aliases:
FAC1, FALZ, NURF301
Chromosome Location:
Chr.17: 67825517 - 67984378 on Build GRCh38
UniGene ID:
Hs.444200
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s5011
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BPTF
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_004459.6
NP_004450.3
13
4365
NM_182641.3
NP_872579.2
11
3987
XM_005257150.3
XP_005257207.1
13
4512
XM_005257151.3
XP_005257208.1
13
4509
XM_005257152.2
XP_005257209.1
13
4512
XM_005257153.3
XP_005257210.1
13
4512
XM_005257154.3
XP_005257211.1
13
4512
XM_005257155.3
XP_005257212.1
12
4323
XM_005257156.3
XP_005257213.1
12
4323
XM_005257157.3
XP_005257214.1
12
4320
XM_005257158.3
XP_005257215.1
11
4134
XM_005257159.2
XP_005257216.1
13
4512
XM_005257160.2
XP_005257217.1
13
4512
XM_005257161.3
XP_005257218.1
13
4512
XM_011524520.2
XP_011522822.1
13
4509
XM_011524521.2
XP_011522823.1
12
4320
XM_011524522.2
XP_011522824.1
11
4131
XM_011524523.2
XP_011522825.1
11
4131
XM_011524524.2
XP_011522826.1
5
1473
XM_011524525.2
XP_011522827.1
11
4131
XM_011524526.2
XP_011522828.1
11
4131
XM_017024353.1
XP_016879842.1
13
4512
XM_017024354.1
XP_016879843.1
7
3241
GenBank mRNA
AB032251.1
-
11
3980
AY282495.1
-
13
3887
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
embryonic placenta development
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
endoderm development
response to wounding
anterior/posterior pattern specification
covalent chromatin modification
positive regulation of transcription, DNA-templated
cellular response to nerve growth factor stimulus
protein binding
DNA-dependent ATPase activity
transcription factor binding
zinc ion binding
sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-xql29:80/10.239.165.153:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0