Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000214.2 | NP_000205.1 | 26 | 4101 | ||
XM_017027707.1 | XP_016883196.1 | 25 | 4206 | |||
GenBank mRNA | AF003837.1 | - | 26 | 4044 | ||
AF028593.1 | - | 26 | 3848 | |||
AK302554.1 | - | 24 | 3451 | |||
AK312722.1 | - | 26 | 3670 | |||
BC098393.1 | - | 26 | 4004 | 30528888 | ||
BC126205.1 | - | 27 | 3609 | 8991921 | ||
BC126207.1 | - | 26 | 3837 | 8991923 | ||
U61276.1 | - | 26 | 3955 | |||
U73936.1 | - | 26 | 3998 | |||
U77720.1 | - | 26 | 3548 |