Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNGA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079878.1 | NP_001073347.1 | 7 | 1714 | ||
NM_001298.2 | NP_001289.1 | 8 | 1768 | |||
XM_006712243.2 | XP_006712306.1 | 8 | 1846 | |||
XM_011510554.2 | XP_011508856.1 | 9 | 1990 | |||
GenBank mRNA | AF065314.1 | - | 7 | 1390 | ||
AK131300.1 | - | 8 | 1504 | |||
AK308149.1 | - | 7 | 1377 | |||
BC096298.1 | - | 7 | 1377 | 40007383 | ||
BC096299.1 | - | 6 | 1323 | 40007387 | ||
BC096300.1 | - | 7 | 1377 | 40007389 |