Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB40AL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031834.1 | NP_001027004.1 | 1 | 543 | ||
GenBank mRNA | BC101169.1 | - | 1 | 543 | 40016064 | |
BC101170.2 | - | 1 | 543 | 40016065 | ||
BC101171.1 | - | 1 | 543 | 40016067 | ||
BC101172.1 | - | 1 | 543 | 40016069 |
Gene Symbol: RAB40A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080879.2 | NP_543155.2 | 3 | 838 | ||
XM_005262083.4 | XP_005262140.1 | 2 | 1315 | |||
XM_011530872.2 | XP_011529174.1 | 2 | 1312 | |||
GenBank mRNA | AB232637.1 | - | 1 | 496 | ||
AF132748.1 | - | 1 | 561 | |||
AF422143.1 | - | 3 | 838 | |||
AK056859.1 | - | 1 | 2615 | |||
BC074854.2 | - | 1 | 527 | 30915461 | ||
BC074855.2 | - | 1 | 527 | 30915224 | ||
BC113501.1 | - | 1 | 579 | 8322553 | ||
BC117232.1 | - | 1 | 579 | 40125783 |