Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000257.3 | NP_000248.2 | 9, 10 | 930 | ||
XM_017021340.1 | XP_016876829.1 | 8, 9 | 865 | |||
GenBank mRNA | AK307590.1 | - | 9, 10 | 904 | ||
BC112171.1 | - | 8, 9 | 854 | 8327639 | ||
BC112173.1 | - | 8, 9 | 854 | 8327641 | ||
EF560725.1 | - | 7, 8 | 797 | |||
EU747717.1 | - | 9, 10 | 905 | |||
M58018.1 | - | 9, 10 | 885 |