Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152778.2 | NP_689991.1 | 12 | 1278 | ||
XM_005262893.1 | XP_005262950.1 | 11 | 1192 | |||
XM_005262896.1 | XP_005262953.1 | 10 | 1045 | |||
XM_005262897.2 | XP_005262954.1 | 9 | 991 | |||
XM_005262898.2 | XP_005262955.1 | 10 | 1088 | |||
XM_011531830.1 | XP_011530132.1 | 10 | 1078 | |||
XM_011531831.1 | XP_011530133.1 | 8 | 877 | |||
XM_011531832.1 | XP_011530134.1 | 9 | 974 | |||
XM_017007989.1 | XP_016863478.1 | 8 | 887 | |||
GenBank mRNA | AK074564.1 | - | 11 | 1127 | ||
AK294439.1 | - | 9 | 906 | |||
AK294957.1 | - | 11 | 1120 | |||
AK315596.1 | - | 11 | 1124 | |||
BC029503.1 | - | 12 | 1243 | 5272664 |