Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPM1J | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005167.5 | NP_005158.5 | 9 | 1328 | ||
GenBank mRNA | AB451327.1 | - | 8 | 683 | ||
AB451471.1 | - | 8 | 683 | |||
AK093270.1 | - | 9 | 1308 | |||
AL133644.1 | - | 10 | 2092 | |||
BC011803.2 | - | 9 | 1252 | |||
BC064628.1 | - | 9 | 1348 | |||
BC073828.1 | - | 9 | 1320 | 5244363 |