Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM95 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320435.1 | NP_001307364.1 | 2, 3 | 292 | ||
NM_001320436.1 | NP_001307365.1 | 2, 3 | 292 | |||
NM_198154.2 | NP_937797.1 | 2, 3 | 292 | |||
XM_017024565.1 | XP_016880054.1 | 2, 3 | 314 | |||
XM_017024568.1 | XP_016880057.1 | 2, 3 | 314 | |||
XM_017024569.1 | XP_016880058.1 | 2, 3 | 314 | |||
XM_017024571.1 | XP_016880060.1 | 2, 3 | 460 | |||
GenBank mRNA | AY358218.1 | - | 2, 3 | 250 | ||
BC040900.1 | - | 2, 3 | 292 | |||
BC107110.2 | - | 2, 3 | 250 | 40022149 |