Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012756.2 | NP_001012774.1 | 4 | 972 | ||
NM_001166036.1 | NP_001159508.1 | 4 | 966 | |||
NM_001166037.1 | NP_001159509.1 | 3 | 858 | |||
NM_001166038.1 | NP_001159510.1 | 3 | 839 | |||
XM_011526897.2 | XP_011525199.1 | 2 | 781 | |||
XM_017026741.1 | XP_016882230.1 | 2 | 1286 | |||
GenBank mRNA | AK291442.1 | - | 3 | 784 | ||
BC042676.1 | - | 4 | 894 | 4821139 | ||
BC119003.1 | - | 3 | 764 | 40082016 | ||
BC122549.1 | - | 2 | 637 | 40082014 | ||
BQ938102.1 | - | 3 | 742 |