Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198493.2 | NP_940895.1 | 3 | 517 | ||
XM_011509466.2 | XP_011507768.1 | 4 | 964 | |||
XM_011509467.1 | XP_011507769.1 | 5 | 1488 | |||
XM_011509468.1 | XP_011507770.1 | 4 | 1456 | |||
XM_011509470.1 | XP_011507772.1 | 3 | 1172 | |||
XM_011509471.2 | XP_011507773.1 | 4 | 2156 | |||
XM_017001123.1 | XP_016856612.1 | 5 | 826 | |||
XM_017001124.1 | XP_016856613.1 | 4 | 2156 | |||
GenBank mRNA | AK127170.1 | - | 3 | 517 | ||
BC126353.1 | - | 3 | 517 | 8992069 | ||
BC126355.1 | - | 3 | 517 | 8992071 |