Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KLHL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198317.2 | NP_938073.1 | 5 | 894 | ||
XM_006710600.3 | XP_006710663.1 | 5 | 1447 | |||
XM_006710601.3 | XP_006710664.1 | 5 | 1447 | |||
GenBank mRNA | AK127585.1 | - | 4 | 541 | ||
AK293258.1 | - | 1 | 23 | |||
AY423763.1 | - | 5 | 894 | |||
BC045768.1 | - | 2 | 907 | |||
BC105742.1 | - | 2 | 410 |