Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BRINP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317188.1 | NP_001304117.1 | 7 | 1535 | ||
NM_199051.2 | NP_950252.1 | 8 | 1930 | |||
XM_011509472.2 | XP_011507774.1 | 9 | 1932 | |||
XM_011509475.2 | XP_011507777.1 | 8 | 1670 | |||
XM_011509476.2 | XP_011507778.1 | 7 | 1748 | |||
XM_017001125.1 | XP_016856614.1 | 10 | 2041 | |||
XM_017001126.1 | XP_016856615.1 | 9 | 1906 | |||
XM_017001127.1 | XP_016856616.1 | 9 | 2070 | |||
XM_017001128.1 | XP_016856617.1 | 9 | 3158 | |||
XM_017001129.1 | XP_016856618.1 | 7 | 1739 | |||
XM_017001130.1 | XP_016856619.1 | 8 | 1715 | |||
XM_017001131.1 | XP_016856620.1 | 9 | 2020 | |||
GenBank mRNA | AB111893.1 | - | 8 | 1677 | ||
AK123024.1 | - | 8 | 1620 | |||
AK123637.1 | - | 8 | 1675 | |||
AK294365.1 | - | 7 | 1337 | |||
BC105052.1 | - | 7 | 1501 | 8144055 | ||
BC105054.1 | - | 7 | 1501 | 8144057 |