Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183062.2 | NP_898885.1 | 2 | 268 | ||
XM_011544175.2 | XP_011542477.1 | 2 | 277 | |||
XM_011544176.2 | XP_011542478.1 | 2 | 277 | |||
GenBank mRNA | BC130400.1 | - | 2 | 340 | 40146431 |