Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145636.1 | NP_001139108.1 | 3, 4 | 454 | ||
XM_006710603.2 | XP_006710666.1 | 4, 5 | 734 | |||
XM_006710604.2 | XP_006710667.1 | 4, 5 | 722 | |||
XM_011541340.1 | XP_011539642.1 | 5, 6 | 858 | |||
XM_011541341.1 | XP_011539643.1 | 3, 4 | 470 | |||
XM_011541343.1 | XP_011539645.1 | 4, 5 | 594 | |||
XM_011541345.1 | XP_011539647.1 | 4, 5 | 668 | |||
XM_011541349.1 | XP_011539651.1 | 3, 4 | 404 | |||
XM_017001140.1 | XP_016856629.1 | 5, 6 | 858 | |||
XM_017001141.1 | XP_016856630.1 | 5, 6 | 858 | |||
XM_017001142.1 | XP_016856631.1 | 3, 4 | 470 | |||
XM_017001143.1 | XP_016856632.1 | 3, 4 | 470 | |||
XM_017001144.1 | XP_016856633.1 | 3, 4 | 477 | |||
XM_017001145.1 | XP_016856634.1 | 3, 4 | 480 | |||
XM_017001146.1 | XP_016856635.1 | 3, 4 | 479 | |||
XM_017001147.1 | XP_016856636.1 | 3, 4 | 477 | |||
GenBank mRNA | AY254217.1 | - | 1, 2 | 74 | ||
BC026115.1 | - | 3, 4 | 458 | 4822241 |