Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1orf228 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145636.1 | NP_001139108.1 | 5 | 725 | ||
XM_006710603.2 | XP_006710666.1 | 6 | 1005 | |||
XM_006710604.2 | XP_006710667.1 | 6 | 993 | |||
XM_011541340.1 | XP_011539642.1 | 7 | 1129 | |||
XM_011541341.1 | XP_011539643.1 | 5 | 741 | |||
XM_011541343.1 | XP_011539645.1 | 6 | 865 | |||
XM_011541345.1 | XP_011539647.1 | 6 | 939 | |||
XM_011541349.1 | XP_011539651.1 | 5 | 675 | |||
XM_011541354.1 | XP_011539656.1 | 4 | 643 | |||
XM_011541355.1 | XP_011539657.1 | 3 | 410 | |||
XM_011541356.2 | XP_011539658.1 | 2 | 160 | |||
XM_017001140.1 | XP_016856629.1 | 7 | 1129 | |||
XM_017001141.1 | XP_016856630.1 | 7 | 1129 | |||
XM_017001142.1 | XP_016856631.1 | 5 | 741 | |||
XM_017001143.1 | XP_016856632.1 | 5 | 741 | |||
XM_017001144.1 | XP_016856633.1 | 5 | 748 | |||
XM_017001145.1 | XP_016856634.1 | 5 | 751 | |||
XM_017001146.1 | XP_016856635.1 | 5 | 750 | |||
XM_017001147.1 | XP_016856636.1 | 5 | 748 | |||
XM_017001148.1 | XP_016856637.1 | 3 | 476 | |||
GenBank mRNA | AY254217.1 | - | 3 | 345 | ||
BC026115.1 | - | 5 | 729 | 4822241 |