Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: CCDC158 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042784.1 | NP_001036249.1 | 22 | 3183 | ||
XM_005262974.4 | XP_005263031.1 | 23 | 3310 | |||
XM_006714204.3 | XP_006714267.1 | 23 | 3322 | |||
XM_011531908.2 | XP_011530210.1 | 24 | 3426 | |||
XM_011531909.2 | XP_011530211.1 | 24 | 3442 | |||
XM_011531910.2 | XP_011530212.1 | 23 | 3903 | |||
XM_011531911.1 | XP_011530213.1 | 23 | 3286 | |||
XM_011531912.2 | XP_011530214.1 | 23 | 3182 | |||
XM_011531913.1 | XP_011530215.1 | 22 | 3210 | |||
XM_011531914.2 | XP_011530216.1 | 22 | 3279 | |||
XM_011531915.2 | XP_011530217.1 | 22 | 3170 | |||
XM_011531916.2 | XP_011530218.1 | 22 | 3104 | |||
XM_011531917.1 | XP_011530219.1 | 23 | 3147 | |||
XM_017008160.1 | XP_016863649.1 | 24 | 3936 | |||
XM_017008161.1 | XP_016863650.1 | 18 | 2387 | |||
GenBank mRNA | AK098600.1 | - | 11 | 1200 | ||
AK098636.1 | - | 12 | 1250 | |||
AL832445.1 | - | 11 | 2913 | |||
BC086869.1 | - | 18 | 2553 |