Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002273.2 | NP_001002273.1 | 7 | 1508 | ||
NM_001002274.2 | NP_001002274.1 | 7 | 1511 | |||
NM_001002275.2 | NP_001002275.1 | 8 | 1565 | |||
NM_001190828.1 | NP_001177757.1 | 7 | 1547 | |||
NM_004001.4 | NP_003992.3 | 8 | 1568 | |||
XM_017000671.1 | XP_016856160.1 | 9 | 1532 | |||
XM_017000672.1 | XP_016856161.1 | 8 | 1475 | |||
GenBank mRNA | AI692546.1 | - | 1 | 54 | ||
AK308150.1 | - | 7 | 1438 |