Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR10A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004460.1 | NP_001004460.1 | 1 | 101 | ||
GenBank mRNA | BC136955.1 | - | 1 | 163 | 9020947 |
Gene Symbol: OR10A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178168.1 | NP_835462.1 | 1 | 143 | ||
GenBank mRNA | AF324499.1 | - | 1 | 143 | ||
BC093651.1 | - | 1 | 174 | 7939496 | ||
BC093653.1 | - | 1 | 174 | 7939498 |