Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FES | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143783.1 | NP_001137255.1 | 11 | 1492 | ||
NM_001143784.1 | NP_001137256.1 | 11 | 1456 | |||
NM_001143785.1 | NP_001137257.1 | 11 | 1401 | |||
NM_002005.3 | NP_001996.1 | 13 | 1762 | |||
XM_005254880.1 | XP_005254937.1 | 12 | 1577 | |||
XM_005254882.1 | XP_005254939.1 | 12 | 1568 | |||
XM_017022005.1 | XP_016877494.1 | 13 | 1746 | |||
XM_017022006.1 | XP_016877495.1 | 12 | 1619 | |||
XM_017022007.1 | XP_016877496.1 | 12 | 1536 | |||
XM_017022008.1 | XP_016877497.1 | 11 | 1410 | |||
XM_017022009.1 | XP_016877498.1 | 13 | 1843 | |||
XM_017022010.1 | XP_016877499.1 | 12 | 1694 | |||
GenBank mRNA | AK300595.1 | - | 11 | 1391 | ||
AK300605.1 | - | 13 | 1720 | |||
AK312545.1 | - | 13 | 1751 | |||
AY513654.1 | - | 10 | 1282 | |||
AY513655.1 | - | 10 | 1282 | |||
AY513656.1 | - | 11 | 1492 | |||
AY513657.1 | - | 11 | 1456 | |||
BC035357.1 | - | 13 | 1738 | 5170548 | ||
X52192.1 | - | 13 | 1741 |