Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: EYS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142800.1 | NP_001136272.1 | 4 | 716 | ||
NM_001142801.1 | NP_001136273.1 | 4 | 716 | |||
NM_001292009.1 | NP_001278938.1 | 4 | 716 | |||
NM_198283.1 | NP_938024.1 | 3 | 575 | |||
GenBank mRNA | AB593112.1 | - | 4 | 672 | ||
AB593114.1 | - | 4 | 681 | |||
AK310478.1 | - | 6 | 748 | |||
AY358133.1 | - | 3 | 575 | |||
BC133011.1 | - | 2 | 440 | |||
BC133013.1 | - | 2 | 437 | |||
BX537914.1 | - | 4 | 716 | |||
BX647741.1 | - | 4 | 706 | |||
FJ416331.1 | - | 4 | 724 | |||
FM209056.1 | - | 4 | 668 |