Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184741.1 | NP_001171670.1 | 5 | 670 | ||
NM_005141.4 | NP_005132.2 | 5 | 847 | |||
GenBank mRNA | AK303788.1 | - | 6 | 765 | ||
AK309592.1 | - | 5 | 792 | |||
AK312972.1 | - | 5 | 847 | |||
BC036007.1 | - | 5 | 638 | |||
BC070245.1 | - | 5 | 808 | |||
BC106760.2 | - | 5 | 816 | 40024790 | ||
BC107766.1 | - | 5 | 816 | 4734415 | ||
EU668333.1 | - | 5 | 833 | |||
J00129.1 | - | 5 | 784 |