Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STRA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182489.1 | NP_872295.1 | 3 | 284 | ||
XM_011516137.2 | XP_011514439.1 | 3 | 924 | |||
XM_011516138.2 | XP_011514440.1 | 3 | 309 | |||
XM_011516139.2 | XP_011514441.1 | 4 | 553 | |||
XM_017012150.1 | XP_016867639.1 | 3 | 924 | |||
GenBank mRNA | AF513502.1 | - | 3 | 284 |