Search Thermo Fisher Scientific
Gene Symbol: FGF5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291812.1 | NP_001278741.1 | 3 | 1084 | ||
NM_004464.3 | NP_004455.2 | 3 | 1372 | |||
NM_033143.2 | NP_149134.1 | 2 | 1268 | |||
GenBank mRNA | AF171928.1 | - | 2 | 1181 | ||
AF535149.1 | - | 3 | 1297 | |||
AK090557.1 | - | 3 | 1084 | |||
BC131502.1 | - | 3 | 1273 | 40082281 |