Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: TBC1D26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178571.4 | NP_848666.2 | 7 | 544 | ||
GenBank mRNA | AB449913.1 | - | 5 | 294 | ||
AK292544.1 | - | 7 | 539 | |||
AK315847.1 | - | 7 | 539 | |||
BC028414.1 | - | 5 | 1218 | |||
BC047400.1 | - | 7 | 538 | 5297645 |