Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181727.1 | NP_859078.1 | 2 | 803 | ||
XM_011533676.2 | XP_011531978.1 | 4 | 1349 | |||
GenBank mRNA | AY221117.1 | - | 2 | 803 | ||
BC033337.1 | - | 2 | 578 | 4838903 | ||
BC126274.1 | - | 2 | 732 | 8991990 | ||
BC136498.1 | - | 2 | 732 | 9020487 |