Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF233 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001207005.1 | NP_001193934.1 | 5 | 923 | ||
NM_181756.2 | NP_861421.2 | 5 | 953 | |||
XM_017026758.1 | XP_016882247.1 | 5 | 1325 | |||
XM_017026759.1 | XP_016882248.1 | 5 | 1106 | |||
GenBank mRNA | AK055310.1 | - | 5 | 1106 | ||
AK095351.1 | - | 5 | 944 | |||
BC136734.1 | - | 4 | 838 | 9020724 | ||
BC136735.1 | - | 4 | 838 | 9020725 |