Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194314.2 | NP_919290.2 | 10 | 2691 | ||
NR_135153.1 | 11 | 2775 | ||||
GenBank mRNA | AK128549.1 | - | 11 | 2754 | ||
AY336933.1 | - | 7 | 1162 | |||
BC132818.1 | - | 10 | 2655 | 40146840 | ||
BC136600.1 | - | 10 | 2655 | 9020590 | ||
BX640886.1 | - | 1 | 396 |