Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303460.1 | NP_001290389.1 | 4 | 334 | ||
NM_004112.3 | NP_004103.1 | 4 | 1026 | |||
NR_130156.1 | 4 | 497 | ||||
GenBank mRNA | AB209390.1 | - | 3 | 2204 | ||
AK293131.1 | - | 4 | 334 | |||
AK308477.1 | - | 5 | 540 | |||
AK308812.1 | - | 4 | 497 | |||
BC032502.1 | - | 4 | 683 | 5242670 | ||
BC108265.1 | - | 4 | 1026 | 4804221 | ||
U66199.1 | - | 4 | 432 | |||
Z70275.1 | - | 3 | 213 |