Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SPEM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199339.2 | NP_955371.2 | 3 | 244 | ||
GenBank mRNA | AK097400.1 | - | 1 | 571 | ||
BC033882.1 | - | 3 | 244 | 5742860 |