Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF568 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204835.1 | NP_001191764.1 | 7 | 1091 | ||
NM_001204836.1 | NP_001191765.1 | 6 | 833 | |||
NM_001204837.1 | NP_001191766.1 | 5 | 774 | |||
NM_198539.3 | NP_940941.2 | 7 | 1094 | |||
XM_017026773.1 | XP_016882262.1 | 8 | 1178 | |||
XM_017026774.1 | XP_016882263.1 | 7 | 783 | |||
XM_017026779.1 | XP_016882268.1 | 7 | 1722 | |||
XM_017026780.1 | XP_016882269.1 | 6 | 1646 | |||
GenBank mRNA | AK093123.1 | - | 6 | 707 | ||
BC031218.1 | - | 7 | 1082 | |||
BC111777.1 | - | 7 | 876 | |||
BC131547.1 | - | 7 | 874 | |||
BX640681.1 | - | 5 | 760 |