Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: WNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184985.1 | NP_001171914.1 | 9 | 3141 | ||
NM_213655.4 | NP_998820.3 | 10 | 3396 | |||
XM_011520997.2 | XP_011519299.1 | 9 | 3512 | |||
XM_011520998.2 | XP_011519300.1 | 9 | 3466 | |||
XM_011520999.2 | XP_011519301.1 | 9 | 3466 | |||
XM_011521000.2 | XP_011519302.1 | 9 | 3466 | |||
XM_011521001.2 | XP_011519303.1 | 9 | 3466 | |||
XM_011521002.2 | XP_011519304.1 | 9 | 3466 | |||
XM_011521003.2 | XP_011519305.1 | 9 | 3466 | |||
XM_011521004.2 | XP_011519306.1 | 9 | 3466 | |||
XM_011521005.2 | XP_011519307.1 | 6 | 1950 | |||
GenBank mRNA | BC130467.1 | - | 1 | 431 | 40146498 | |
BC130469.1 | - | 1 | 431 | 40146500 | ||
BJ997509.1 | - | 1 | 521 | |||
JQ358908.1 | - | 9 | 2498 |