Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159699.1 | NP_001153171.1 | 5 | 903 | ||
NM_001159700.1 | NP_001153172.1 | 6 | 898 | |||
NM_001159701.1 | NP_001153173.1 | 5 | 774 | |||
NM_001159702.2 | NP_001153174.1 | 6 | 967 | |||
NM_001159704.1 | NP_001153176.1 | 5 | 1349 | |||
NM_001167819.1 | NP_001161291.1 | 6 | 835 | |||
NM_001449.4 | NP_001440.2 | 6 | 967 | |||
NR_027621.1 | 6 | 1049 | ||||
XM_006724743.2 | XP_006724806.1 | 5 | 771 | |||
XM_006724744.2 | XP_006724807.1 | 6 | 849 | |||
XM_006724745.3 | XP_006724808.1 | 6 | 787 | |||
XM_006724746.2 | XP_006724809.1 | 5 | 722 | |||
XM_006724747.2 | XP_006724810.1 | 5 | 722 | |||
XM_017029357.1 | XP_016884846.1 | 6 | 849 | |||
GenBank mRNA | AF063002.1 | - | 5 | 722 | ||
AF098518.1 | - | 5 | 691 | |||
AK091702.1 | - | 5 | 465 | |||
AK094024.1 | - | 4 | 483 | |||
AK122708.1 | - | 5 | 1350 | |||
AK223299.1 | - | 4 | 672 | |||
AK289411.1 | - | 6 | 1049 | |||
AK297124.1 | - | 6 | 740 | |||
AK299379.1 | - | 5 | 629 | |||
AK299381.1 | - | 5 | 749 | |||
AK299450.1 | - | 6 | 683 | |||
AK300966.1 | - | 7 | 774 | |||
AK301642.1 | - | 5 | 771 | |||
AK304680.1 | - | 6 | 826 | |||
BC010998.2 | - | 6 | 846 | 4039973 | ||
BC088369.1 | - | 5 | 736 | 30529295 | ||
CR456974.1 | - | 4 | 638 | |||
DA917361.1 | - | 4 | 533 | |||
U29538.1 | - | 4 | 659 |
Gene Symbol: LOC100128164 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027622.1 | 2 | 3902 | |||
GenBank mRNA | AL833309.1 | - | 2 | 3396 |