Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOMM20L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207377.2 | NP_997260.1 | 3 | 226 | ||
XM_011536742.2 | XP_011535044.1 | 3 | 269 | |||
XM_011536743.2 | XP_011535045.1 | 3 | 272 | |||
GenBank mRNA | AY358266.1 | - | 3 | 238 | ||
BC137557.1 | - | 3 | 224 | 9021557 | ||
BC137563.1 | - | 3 | 224 | 9021563 |