Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCL4L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207007.3 | NP_996890.1 | 3 | 653 | ||
NR_111969.1 | 2 | 538 | ||||
GenBank mRNA | AY766447.1 | - | 3 | 498 | ||
AY766449.1 | - | 2 | 383 | |||
BC070310.1 | - | 3 | 495 | 6708338 |
Gene Symbol: CCL4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291468.1 | NP_001278397.1 | 3 | 483 | ||
NM_001291469.1 | NP_001278398.1 | 3 | 687 | |||
NM_001291470.1 | NP_001278399.1 | 2 | 1089 | |||
NM_001291471.1 | NP_001278400.1 | 3 | 667 | |||
NM_001291472.1 | NP_001278401.1 | 3 | 646 | |||
NM_001291473.1 | NP_001278402.1 | 3 | 616 | |||
NM_001291474.1 | NP_001278403.1 | 2 | 572 | |||
NM_001291475.1 | NP_001278404.1 | 3 | 677 | |||
NR_111970.1 | 2 | 368 | ||||
GenBank mRNA | AY766450.1 | - | 3 | 483 | ||
AY766451.1 | - | 3 | 687 | |||
AY766452.1 | - | 2 | 1089 | |||
AY766453.1 | - | 3 | 667 | |||
AY766454.1 | - | 3 | 646 | |||
AY766455.1 | - | 3 | 616 | |||
AY766456.1 | - | 2 | 368 | |||
AY766457.1 | - | 2 | 572 | |||
AY766458.1 | - | 3 | 677 | |||
BC092445.1 | - | 3 | 684 | 30416435 | ||
CD521054.1 | - | 1 | 416 |