Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM12B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203390.2 | NP_976324.2 | 3 | 829 | ||
XM_005250915.4 | XP_005250972.1 | 4 | 906 | |||
XM_011517027.2 | XP_011515329.1 | 3 | 1555 | |||
XM_011517028.2 | XP_011515330.1 | 3 | 850 | |||
XM_011517029.2 | XP_011515331.1 | 2 | 796 | |||
XM_017013406.1 | XP_016868895.1 | 3 | 1089 | |||
XM_017013407.1 | XP_016868896.1 | 5 | 961 | |||
XM_017013408.1 | XP_016868897.1 | 3 | 849 |