Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257360.1 | NP_001244289.1 | 12 | 2085 | ||
NM_001257361.1 | NP_001244290.1 | 11 | 1368 | |||
NM_001308170.1 | NP_001295099.1 | 10 | 1488 | |||
NM_004037.7 | NP_004028.3 | 11 | 1952 | |||
NM_139156.3 | NP_631895.1 | 11 | 1732 | |||
NM_203404.1 | NP_981949.1 | 9 | 1370 | |||
GenBank mRNA | AK025706.1 | - | 11 | 1708 | ||
AK296394.1 | - | 11 | 1324 | |||
AK302939.1 | - | 11 | 1330 | |||
AK308907.1 | - | 12 | 1558 | |||
AK314494.1 | - | 12 | 2008 | |||
BC007711.2 | - | 11 | 1657 | 4101667 | ||
BC075844.1 | - | 11 | 1657 | 4130690 | ||
S47833.1 | - | 10 | 1343 | |||
U16270.1 | - | 6 | 756 |