Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BIN1 SIRNA Assays ›
Gene Symbol:
BIN1
Gene Name:
bridging integrator 1
Gene Aliases:
AMPH2, AMPHL, SH3P9
Chromosome Location:
Chr.2: 127048023 - 127107400 on Build GRCh38
UniGene ID:
Hs.193163
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s526091
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BIN1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320632.1
NP_001307561.1
12, 13
1441
NM_001320633.1
NP_001307562.1
13, 14
1579
NM_001320634.1
NP_001307563.1
11, 12
1162
NM_001320640.1
NP_001307569.1
14, 15
1708
NM_001320641.1
NP_001307570.1
16, 17
1855
NM_001320642.1
NP_001307571.1
17, 18
2442
NM_004305.3
NP_004296.1
13, 14
1531
NM_139343.2
NP_647593.1
17, 18
1948
NM_139344.2
NP_647594.1
16, 17
1819
NM_139345.2
NP_647595.1
14, 15
1687
NM_139346.2
NP_647596.1
14, 15
1660
NM_139347.2
NP_647597.1
14, 15
1723
NM_139348.2
NP_647598.1
13, 14
1615
NM_139349.2
NP_647599.1
13, 14
1594
NM_139350.2
NP_647600.1
12, 13
1486
NM_139351.2
NP_647601.1
11, 12
1396
XM_005263642.3
XP_005263699.1
18, 19
2066
XM_005263643.3
XP_005263700.1
17, 18
1973
XM_005263645.2
XP_005263702.1
17, 18
1457
XM_006712425.3
XP_006712488.1
16, 17
1934
XM_006712426.3
XP_006712489.1
16, 17
1934
XM_006712427.2
XP_006712490.1
16, 17
1412
XM_006712428.3
XP_006712491.1
15, 16
1826
XM_006712429.3
XP_006712492.1
15, 16
1805
XM_006712431.3
XP_006712494.1
14, 15
1736
XM_006712432.3
XP_006712495.1
14, 15
1697
XM_006712433.3
XP_006712496.1
13, 14
1607
XM_006712434.3
XP_006712497.1
12, 13
1562
XM_011510975.2
XP_011509277.1
18, 19
2699
XM_017003819.1
XP_016859308.1
18, 19
1880
XM_017003820.1
XP_016859309.1
14, 15
1796
XM_017003821.1
XP_016859310.1
13, 14
1691
XM_017003822.1
XP_016859311.1
12, 13
1598
XM_017003823.1
XP_016859312.1
13, 14
1151
XM_017003824.1
XP_016859313.1
12, 13
1082
XM_017003825.1
XP_016859314.1
12, 13
1376
XM_017003826.1
XP_016859315.1
12, 13
1043
GenBank mRNA
AF001383.1
-
13, 14
1402
AF004015.1
-
17, 18
1564
AF043898.1
-
14, 15
1339
AF043899.1
-
14, 15
1303
AF043900.1
-
13, 14
1210
AF043901.1
-
16, 17
1435
AF068914.1
-
12, 13
1162
AF068915.1
-
11, 12
1072
AF068916.1
-
12, 13
1117
AF068917.1
-
13, 14
1291
AF068918.1
-
14, 15
1336
AF070576.1
-
13, 14
1170
AK127325.1
-
3, 4
4431
AK295300.1
-
13, 14
1297
AK301153.1
-
11, 12
1162
AL713697.1
-
17, 18
1909
BC004101.2
-
12, 13
1162
3688734
BM805510.1
-
3, 4
286
BT006865.1
-
12, 13
1102
DR761993.1
-
5, 6
555
U68485.1
-
13, 14
1193
U87558.1
-
17, 18
1585
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
endocytosis
nucleus organization
cell proliferation
viral process
regulation of endocytosis
muscle cell differentiation
positive regulation of apoptotic process
regulation of neuron differentiation
positive regulation of astrocyte differentiation
lipid tube assembly
regulation of cell cycle arrest
protein binding
identical protein binding
tau protein binding
actin filament binding
RNA polymerase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7cf5v:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0