Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077490.2 | NP_001070958.1 | 1 | 314 | ||
NM_001309883.1 | NP_001296812.1 | 1 | 314 | |||
NM_080425.3 | NP_536350.2 | 1 | 314 | |||
XM_017027812.1 | XP_016883301.1 | 1 | 581 | |||
XM_017027813.1 | XP_016883302.1 | 1 | 581 | |||
XM_017027814.1 | XP_016883303.1 | 1 | 581 |