Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHEX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000444.5 | NP_000435.3 | 7, 8 | 1418 | ||
NM_001282754.1 | NP_001269683.1 | 7, 8 | 1418 | |||
XM_011545533.1 | XP_011543835.1 | 3, 4 | 227 | |||
XM_017029579.1 | XP_016885068.1 | 3, 4 | 228 | |||
GenBank mRNA | AD000712.1 | - | 7, 8 | 1015 | ||
AK298030.1 | - | 7, 8 | 964 | |||
AK301528.1 | - | 6, 7 | 1154 | |||
BC105057.1 | - | 7, 8 | 940 | 8144060 | ||
BC105059.1 | - | 7, 8 | 940 | 8144062 | ||
U60475.1 | - | 7, 8 | 849 | |||
U75645.1 | - | 7, 8 | 932 | |||
U82970.1 | - | 7, 8 | 1459 | |||
U87284.1 | - | 7, 8 | 1057 |