Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACCSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031854.2 | NP_001027025.2 | 12, 13 | 1537 | ||
XM_017017714.1 | XP_016873203.1 | 15, 16 | 1910 | |||
XM_017017715.1 | XP_016873204.1 | 12, 13 | 1550 | |||
GenBank mRNA | AB231734.1 | - | 12, 13 | 1537 | ||
BC144554.1 | - | 12, 13 | 1550 | |||
DW009984.1 | - | 2, 3 | 236 |