Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR5M1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004740.1 | NP_001004740.1 | 1 | 771 |
Gene Symbol: OR5M10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004741.1 | NP_001004741.1 | 1 | 771 | ||
GenBank mRNA | BC140733.1 | - | 1 | 837 | 9021601 |