Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NT5C2 SIRNA Assays ›
Gene Symbol:
NT5C2
Gene Name:
5'-nucleotidase, cytosolic II
Gene Aliases:
GMP, NT5B, PNT5, SPG45, SPG65, cN-II
Chromosome Location:
Chr.10: 103088017 - 103193306 on Build GRCh38
UniGene ID:
Hs.97439
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s527359
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NT5C2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134373.2
NP_001127845.1
12
971
NM_012229.4
NP_036361.1
13
1069
XM_005269632.4
XP_005269689.1
13
980
XM_005269633.4
XP_005269690.1
14
1073
XM_005269634.4
XP_005269691.1
14
1105
XM_005269635.4
XP_005269692.1
13
1295
XM_005269636.4
XP_005269693.1
13
1081
XM_005269637.4
XP_005269694.1
12
1799
XM_005269638.4
XP_005269695.1
12
3100
XM_005269639.4
XP_005269696.1
11
824
XM_005269640.4
XP_005269697.1
14
1096
XM_005269641.4
XP_005269698.1
13
998
XM_005269642.4
XP_005269699.1
12
923
XM_005269643.4
XP_005269700.1
11
863
XM_005269644.4
XP_005269701.1
15
1120
XM_005269645.4
XP_005269702.1
14
1022
XM_005269646.4
XP_005269703.1
13
948
XM_006717721.3
XP_006717784.1
13
1022
XM_006717723.3
XP_006717786.1
11
819
XM_011539537.2
XP_011537839.1
14
1105
XM_017015947.1
XP_016871436.1
11
849
XM_017015948.1
XP_016871437.1
14
1205
XM_017015949.1
XP_016871438.1
14
1190
XM_017015950.1
XP_016871439.1
12
906
XM_017015951.1
XP_016871440.1
11
832
XM_017015952.1
XP_016871441.1
13
1129
XM_017015953.1
XP_016871442.1
14
1524
XM_017015954.1
XP_016871443.1
13
1463
XM_017015955.1
XP_016871444.1
13
1150
XM_017015956.1
XP_016871445.1
13
1356
XM_017015957.1
XP_016871446.1
15
1214
XM_017015958.1
XP_016871447.1
16
1333
XM_017015959.1
XP_016871448.1
12
924
XM_017015960.1
XP_016871449.1
13
887
XM_017015961.1
XP_016871450.1
13
1282
XM_017015962.1
XP_016871451.1
13
1116
XM_017015963.1
XP_016871452.1
13
1220
XM_017015964.1
XP_016871453.1
13
1199
XM_017015965.1
XP_016871454.1
12
1076
XM_017015966.1
XP_016871455.1
12
1076
XM_017015967.1
XP_016871456.1
12
1011
XM_017015968.1
XP_016871457.1
13
1004
XM_017015969.1
XP_016871458.1
13
1098
XM_017015970.1
XP_016871459.1
11
1741
XM_017015971.1
XP_016871460.1
10
785
XM_017015972.1
XP_016871461.1
14
1039
XM_017015973.1
XP_016871462.1
15
1063
XM_017015974.1
XP_016871463.1
14
1063
XM_017015975.1
XP_016871464.1
12
904
XM_017015976.1
XP_016871465.1
12
910
GenBank mRNA
AK127670.1
-
13
1060
AK291667.1
-
12
937
AK295593.1
-
11
1113
AK312517.1
-
12
937
BC001595.1
-
12
1020
3543103
BX648078.1
-
14
2871
D38524.1
-
12
959
Back To Top
More Information
Panther Classification:
Molecular Function -
nucleotide phosphatase
Gene Ontology Categories:
Function(s)
Process(es)
purine nucleotide catabolic process
phosphorylation
dephosphorylation
drug metabolic process
IMP metabolic process
adenosine metabolic process
nucleotide binding
protein binding
5'-nucleotidase activity
metal ion binding
nucleoside phosphotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-j4kkz:80/10.239.163.216:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0