Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: OR8B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005467.1 | NP_001005467.1 | 1 | 643 | ||
XM_017017716.1 | XP_016873205.1 | 6 | 2009 | |||
XM_017017717.1 | XP_016873206.1 | 6 | 1312 | |||
XM_017017718.1 | XP_016873207.1 | 4 | 1740 |
Gene Symbol: OR8B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005468.1 | NP_001005468.1 | 1 | 643 | ||
XM_017017535.1 | XP_016873024.1 | 3 | 875 | |||
XM_017017536.1 | XP_016873025.1 | 3 | 861 |