Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR8A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005194.1 | NP_001005194.1 | 1 | 666 | ||
GenBank mRNA | AY726592.1 | - | 1 | 542 | ||
BC137194.1 | - | 1 | 738 | 9021191 | ||
BC137197.1 | - | 1 | 738 | 9021194 |