Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OR4K2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005501.1 | NP_001005501.1 | 1 | 248 | ||
GenBank mRNA | BC136941.1 | - | 1 | 284 | 9020933 | |
BC136942.1 | - | 1 | 284 | 9020934 |
Gene Symbol: G3BP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006714749.3 | XP_006714812.1 | 12 | 5865 | ||
XM_006714750.3 | XP_006714813.1 | 12 | 5804 | |||
XM_017008922.1 | XP_016864411.1 | 12 | 6151 | |||
XM_017008923.1 | XP_016864412.1 | 12 | 6120 | |||
GenBank mRNA | AK128316.1 | - | 1 | 1642 |