Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014244.4 | NP_055059.2 | 16 | 2425 | ||
XM_017010063.1 | XP_016865552.1 | 15 | 1861 | |||
GenBank mRNA | AJ003125.1 | - | 16 | 2323 | ||
CO247372.1 | - | 2 | 58 |