Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ACADM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000016.5 | NP_000007.1 | 11, 12 | 1643 | ||
NM_001127328.2 | NP_001120800.1 | 11, 12 | 1655 | |||
NM_001286042.1 | NP_001272971.1 | 10, 11 | 1555 | |||
NM_001286043.1 | NP_001272972.1 | 12, 13 | 1742 | |||
NM_001286044.1 | NP_001272973.1 | 8, 9 | 1373 | |||
GenBank mRNA | AF251043.1 | - | 11, 12 | 1222 | ||
AK296045.1 | - | 8, 9 | 953 | |||
AK301005.1 | - | 8, 9 | 1111 | |||
AK301717.1 | - | 10, 11 | 1147 | |||
AK312629.1 | - | 11, 12 | 1233 | |||
AK315946.1 | - | 10, 11 | 1196 | |||
BC005377.1 | - | 11, 12 | 1237 | 3934299 | ||
BX647734.1 | - | 12, 13 | 1381 | |||
KU177876.1 | - | 11, 12 | 1202 | |||
KU177877.1 | - | 10, 11 | 1114 | |||
M16827.1 | - | 11, 12 | 1220 |