Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MID1 SIRNA Assays ›
Gene Symbol:
MID1
Gene Name:
midline 1
Gene Aliases:
BBBG1, FXY, GBBB1, MIDIN, OGS1, OS, OSX, RNF59, TRIM18, XPRF, ZNFXY
Chromosome Location:
Chr.X: 10445310 - 11111264 on Build GRCh38
UniGene ID:
Hs.27695
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s529207
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MID1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000381.3
NP_000372.1
3
1144
NM_001098624.2
NP_001092094.1
3
1074
NM_001193277.1
NP_001180206.1
3
933
NM_001193278.1
NP_001180207.1
2
799
NM_001193279.1
NP_001180208.1
2
799
NM_001193280.1
NP_001180209.1
2
685
NM_001193281.1
NP_001180210.1
2
685
NM_033289.1
NP_150631.1
3
1030
NM_033290.3
NP_150632.1
3
936
XM_005274536.1
XP_005274593.1
3
956
XM_005274537.1
XP_005274594.1
3
933
XM_006724492.3
XP_006724555.1
3
1555
XM_011545525.2
XP_011543827.1
5
1257
XM_011545526.2
XP_011543828.1
5
1292
XM_011545527.2
XP_011543829.1
3
1202
XM_017029534.1
XP_016885023.1
3
1122
XM_017029535.1
XP_016885024.1
5
1143
XM_017029536.1
XP_016885025.1
5
1372
XM_017029537.1
XP_016885026.1
5
1143
XM_017029538.1
XP_016885027.1
3
842
XM_017029539.1
XP_016885028.1
3
1004
XM_017029540.1
XP_016885029.1
5
1257
XM_017029541.1
XP_016885030.1
5
1257
XM_017029542.1
XP_016885031.1
5
1257
GenBank mRNA
AF035360.1
-
3
830
AF041206.1
-
3
1024
AF041208.1
-
3
1144
AF041210.1
-
2
633
AF230976.1
-
3
929
AF230977.1
-
3
929
AF269101.1
-
4
1052
AK025187.1
-
2
115
AK226104.1
-
3
848
AK291215.1
-
3
1026
AK297205.1
-
4
940
AK299061.1
-
3
969
AK315095.1
-
3
975
AY539962.1
-
2
126
AY539963.1
-
3
287
AY539964.1
-
2
126
AY539965.1
-
2
126
AY539976.1
-
2
212
AY539988.1
-
2
629
AY539989.1
-
2
358
AY539990.1
-
3
610
AY539994.1
-
2
743
AY539995.1
-
2
358
AY539996.1
-
2
244
AY540001.1
-
2
358
AY540002.1
-
2
244
AY540014.1
-
2
244
AY540015.1
-
2
358
AY540016.1
-
2
358
AY540022.1
-
2
743
AY540023.1
-
2
629
BC053626.1
-
3
1001
6163881
Y13667.1
-
3
929
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
microtubule cytoskeleton organization
negative regulation of microtubule depolymerization
pattern specification process
positive regulation of stress-activated MAPK cascade
protein localization to microtubule
interferon-gamma-mediated signaling pathway
protein binding
microtubule binding
zinc ion binding
ligase activity
ubiquitin protein ligase binding
identical protein binding
protein homodimerization activity
protein heterodimerization activity
phosphoprotein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-tbml7:80/10.239.166.229:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0