Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000274.3 | NP_000265.1 | 6 | 862 | ||
NM_001171814.1 | NP_001165285.1 | 5 | 634 | |||
NM_001322965.1 | NP_001309894.1 | 6 | 857 | |||
NM_001322966.1 | NP_001309895.1 | 7 | 1177 | |||
NM_001322967.1 | NP_001309896.1 | 7 | 982 | |||
NM_001322968.1 | NP_001309897.1 | 8 | 1069 | |||
NM_001322969.1 | NP_001309898.1 | 7 | 949 | |||
NM_001322970.1 | NP_001309899.1 | 8 | 1165 | |||
NM_001322971.1 | NP_001309900.1 | 4 | 541 | |||
NM_001322974.1 | NP_001309903.1 | 6 | 748 | |||
XM_017016279.1 | XP_016871768.1 | 6 | 2657 | |||
GenBank mRNA | AB208817.1 | - | 6 | 2647 | ||
AK296032.1 | - | 5 | 581 | |||
AK312561.1 | - | 6 | 809 | |||
AK315947.1 | - | 5 | 581 | |||
BC000964.2 | - | 6 | 790 | 3449883 | ||
BC016928.1 | - | 6 | 794 | 3913352 | ||
CD683561.1 | - | 4 | 510 | |||
CR457045.1 | - | 5 | 729 | |||
CR749808.1 | - | 5 | 584 | |||
M12267.1 | - | 6 | 783 | |||
M14963.1 | - | 5 | 773 | |||
M23204.1 | - | 6 | 780 | |||
Y07511.1 | - | 5 | 769 |