Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000274.3 | NP_000265.1 | 4 | 624 | ||
NM_001171814.1 | NP_001165285.1 | 3 | 396 | |||
NM_001322965.1 | NP_001309894.1 | 4 | 619 | |||
NM_001322966.1 | NP_001309895.1 | 5 | 939 | |||
NM_001322967.1 | NP_001309896.1 | 5 | 744 | |||
NM_001322968.1 | NP_001309897.1 | 6 | 831 | |||
NM_001322969.1 | NP_001309898.1 | 5 | 711 | |||
NM_001322970.1 | NP_001309899.1 | 6 | 927 | |||
NM_001322974.1 | NP_001309903.1 | 3 | 396 | |||
XM_017016279.1 | XP_016871768.1 | 4 | 566 | |||
GenBank mRNA | AB208817.1 | - | 4 | 555 | ||
AK296032.1 | - | 3 | 343 | |||
AK312561.1 | - | 4 | 571 | |||
AK315947.1 | - | 3 | 343 | |||
BC000964.2 | - | 4 | 552 | 3449883 | ||
BC016928.1 | - | 4 | 556 | 3913352 | ||
CD104356.1 | - | 4 | 567 | |||
CR457045.1 | - | 3 | 491 | |||
CR749808.1 | - | 3 | 346 | |||
M12267.1 | - | 4 | 545 | |||
M14963.1 | - | 3 | 535 | |||
M23204.1 | - | 4 | 542 | |||
Y07511.1 | - | 3 | 531 |